Assay Details
Gene Symbol: HTR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048119.1 | NP_001041584.1 | 866 | 112 | ||
XM_014113197.2 | XP_013968672.1 | 1064 | 112 | |||
GenBank mRNA | DQ483094.1 | - | 866 | 112 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
