Assay Details
Gene Symbol: MIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313795.1 | NP_001300724.1 | 560 | 81 | ||
NR_132651.1 | 560 | 81 | ||||
XM_022424010.1 | XP_022279718.1 | 1414 | 81 | |||
GenBank mRNA | DN863767.1 | - | 560 | 81 | ||
DN864176.1 | - | 308 | 81 | |||
EF191197.1 | - | 560 | 81 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
