Assay Details
Gene Symbol: GPX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164454.1 | NP_001157926.1 | 1 - 2 | 145 | 85 | |
GenBank mRNA | DN374277.1 | - | 1 - 2 | 143 | 85 | |
DN439024.1 | - | 1 - 2 | 155 | 85 | ||
DN904267.1 | - | 1 - 2 | 138 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
