Assay Details
Gene Symbol: PRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197182.1 | NP_001184111.1 | 1 - 2 | 542 | 74 | |
XM_005618879.2 | XP_005618936.1 | 2 - 3 | 670 | 74 | ||
GenBank mRNA | FJ973622.1 | - | 1 - 2 | 538 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
