Assay Details
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003242.2 | NP_001003242.2 | 1 - 2 | 148 | 73 | |
GenBank mRNA | DN349868.1 | - | 1 - 2 | 148 | 73 | |
DN392295.1 | - | 1 - 2 | 99 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
