Assay Details
Gene Symbol: SAP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252161.1 | NP_001239090.1 | 282 | 104 | ||
XM_014107336.2 | XP_013962811.1 | 850 | 104 | |||
GenBank mRNA | CO673277.1 | - | 370 | 104 | ||
CX993913.1 | - | 356 | 104 | |||
DT542607.1 | - | 261 | 104 |

More Information
Set Membership: |
![]() ![]() |
