Assay Details
Gene Symbol: RPS27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252228.1 | NP_001239157.1 | 4 - 5 | 227 | 69 | |
GenBank mRNA | GR889329.1 | - | 4 - 5 | 227 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
