Assay Details
Gene Symbol: GPX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252323.1 | NP_001239252.1 | 1 - 2 | 205 | 79 | |
GenBank mRNA | DN379330.1 | - | 1 - 2 | 203 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
