Assay Details
Gene Symbol: RPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253906.1 | NP_001240835.1 | 3 - 4 | 288 | 91 | |
NM_001253907.1 | NP_001240836.1 | 3 - 4 | 285 | 91 | ||
GenBank mRNA | GR885819.1 | - | 3 - 4 | 285 | 91 | |
GR899925.1 | - | 3 - 4 | 282 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
