Assay Details
Gene Symbol: RPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253906.1 | NP_001240835.1 | 4 - 5 | 422 | 85 | |
NM_001253907.1 | NP_001240836.1 | 4 - 5 | 419 | 85 | ||
GenBank mRNA | GR885819.1 | - | 4 - 5 | 419 | 85 | |
GR899925.1 | - | 4 - 5 | 416 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
