Assay Details
Gene Symbol: RPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253906.1 | NP_001240835.1 | 5 - 6 | 522 | 86 | |
NM_001253907.1 | NP_001240836.1 | 5 - 6 | 519 | 86 | ||
GenBank mRNA | GR885819.1 | - | 5 - 6 | 519 | 86 | |
GR899925.1 | - | 5 - 6 | 516 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
