Assay Details
Gene Symbol: HOTAIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131937.1 | 2 - 3 | 339 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeobox C11 | HOXC11 | -8.8kb | Anti-sense | Chr. 27 - 1287326 - 1289577 on NCBI Build CanFam3.1 | |
Downstream | homeobox C12 | HOXC12 | 5.3kb | Anti-sense | Chr. 27 - 1305219 - 1307350 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
