Assay Details
Gene Symbol: ABO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284436.1 | NP_001271365.1 | 3 - 4 | 157 | 74 | |
GenBank mRNA | AB638847.1 | - | 3 - 4 | 157 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
