Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286869.1 | NP_001273798.1 | 1 | 275 | 81 | |
NM_001286870.1 | NP_001273799.1 | 1 | 263 | 81 | ||
GenBank mRNA | DN269563.1 | - | 237 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
