Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286869.1 | NP_001273798.1 | 1 | 507 | 70 | |
NM_001286870.1 | NP_001273799.1 | 1 | 495 | 70 | ||
GenBank mRNA | AB578924.1 | - | 256 | 70 | ||
AB578925.1 | - | 256 | 70 | |||
DN269563.1 | - | 469 | 70 | |||
DT539293.1 | - | 78 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
