Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286870.1 | NP_001273799.1 | 75 | 55 | ||
GenBank mRNA | AB578930.1 | - | 66 | 55 | ||
AB578931.1 | - | 70 | 55 | |||
AB578932.1 | - | 35 | 55 | |||
DN269563.1 | - | 49 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286870.1 transcripts.
Set Membership: |
![]() ![]() |
