Assay Details
Gene Symbol: CHST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287130.1 | NP_001274059.1 | 2 - 3 | 89 | 75 | |
XM_014113962.2 | XP_013969437.1 | 2 - 3 | 230 | 75 | ||
XM_022418049.1 | XP_022273757.1 | 3 - 4 | 322 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001287130.1, XM_014113962.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
