Assay Details
Gene Symbol: RPL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313766.1 | NP_001300695.1 | 3 - 4 | 252 | 86 | |
XM_005616310.2 | XP_005616367.1 | 4 - 5 | 555 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005616310.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
