Assay Details
Gene Symbol: RPL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313766.1 | NP_001300695.1 | 4 - 5 | 349 | 64 | |
XM_005616310.2 | XP_005616367.1 | 5 - 6 | 652 | 64 | ||
GenBank mRNA | AJ388525.1 | - | 3 - 4 | 249 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
