Assay Details
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003266.2 | NP_001003266.1 | 1 - 2 | 97 | 89 | |
NM_001313790.1 | NP_001300719.1 | 1 - 2 | 97 | 89 | ||
NM_001313791.1 | NP_001300720.1 | 1 - 2 | 97 | 89 | ||
GenBank mRNA | DN347413.1 | - | 1 - 2 | 109 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003266.2, NM_001313790.1, NM_001313791.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
