Assay Details
Gene Symbol: eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002645.3 | NP_001002645.3 | 4 - 5 | 604 | 77 | |
XM_021470136.1 | XP_021325811.1 | 4 - 5 | 515 | 77 | ||
XM_021470137.1 | XP_021325812.1 | 4 - 5 | 594 | 77 | ||
GenBank mRNA | BC076531.1 | - | 4 - 5 | 531 | 77 | |
BC155309.1 | - | 7 - 8 | 748 | 77 | ||
EH435933.1 | - | 4 - 5 | 556 | 77 | ||
EH589679.1 | - | 4 - 5 | 550 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
