Assay Details
Gene Symbol: eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002645.3 | NP_001002645.3 | 6 - 7 | 779 | 131 | |
XM_021470136.1 | XP_021325811.1 | 6 - 7 | 690 | 131 | ||
XM_021470137.1 | XP_021325812.1 | 6 - 7 | 769 | 131 | ||
GenBank mRNA | BC076531.1 | - | 6 - 7 | 131 | ||
BC155309.1 | - | 9 - 10 | 923 | 131 | ||
EH589679.1 | - | 6 - 7 | 725 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
