Assay Details
Gene Symbol: cenpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003555.3 | NP_001003555.2 | 4 - 5 | 459 | 141 | |
GenBank mRNA | BC078335.1 | - | 4 - 5 | 433 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
