Assay Details
Gene Symbol: msna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004296.1 | NP_001004296.1 | 5 - 6 | 616 | 83 | |
XM_009301281.3 | XP_009299556.1 | 5 - 6 | 514 | 83 | ||
GenBank mRNA | BC049479.1 | - | 5 - 6 | 680 | 83 | |
BC081551.1 | - | 5 - 6 | 616 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
