Assay Details
Gene Symbol: mapk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013451.3 | NP_001013469.2 | 5 - 6 | 2192 | 67 | |
GenBank mRNA | DQ360072.1 | - | 5 - 6 | 2192 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
