Assay Details
Gene Symbol: mrps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017590.1 | NP_001017590.1 | 2 - 3 | 336 | 60 | |
GenBank mRNA | BC092991.1 | - | 2 - 3 | 336 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
