Assay Details
Gene Symbol: pgam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007323.2 | NP_001007324.2 | 3 - 4 | 810 | 90 | |
XM_005165077.3 | XP_005165134.1 | 3 - 4 | 804 | 90 | ||
GenBank mRNA | BC085627.1 | - | 3 - 4 | 555 | 90 | |
BC095654.1 | - | 3 - 4 | 761 | 90 | ||
BC164655.1 | - | 3 - 4 | 535 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
