Assay Details
Gene Symbol: mrpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076725.1 | NP_001070193.1 | 3 - 4 | 347 | 102 | |
GenBank mRNA | BC122374.1 | - | 3 - 4 | 347 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
