Assay Details
Gene Symbol: bmi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080751.1 | NP_001074220.1 | 5 - 6 | 422 | 104 | |
XM_021479461.1 | XP_021335136.1 | 5 - 6 | 499 | 104 | ||
GenBank mRNA | BC053151.1 | - | 5 - 6 | 422 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
