Assay Details
Gene Symbol: ctnnbip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131594.1 | NP_571669.1 | 3 - 4 | 306 | 95 | |
GenBank mRNA | AB046866.1 | - | 3 - 4 | 306 | 95 | |
BC054693.1 | - | 3 - 4 | 275 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
