Assay Details
Gene Symbol: tradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131607.2 | NP_571682.1 | 4 - 5 | 569 | 113 | |
GenBank mRNA | BC083425.1 | - | 4 - 5 | 569 | 113 | |
BC164053.1 | - | 3 - 4 | 488 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
