Assay Details
Gene Symbol: gria2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131894.1 | NP_571969.1 | 15 - 16 | 2834 | 63 | |
XM_005170898.3 | XP_005170955.1 | 15 - 16 | 2856 | 63 | ||
GenBank mRNA | AF361752.1 | - | 5 - 6 | 1034 | 63 | |
AF525743.1 | - | 15 - 16 | 2834 | 63 | ||
BC162655.1 | - | 15 - 16 | 2665 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
