Assay Details
Gene Symbol: aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199690.2 | NP_955984.2 | 4 - 5 | 642 | 103 | |
XM_005173083.4 | XP_005173140.1 | 4 - 5 | 684 | 103 | ||
XM_009307693.3 | XP_009305968.1 | 4 - 5 | 687 | 103 | ||
GenBank mRNA | BC059643.1 | - | 4 - 5 | 640 | 103 | |
BC065447.1 | - | 4 - 5 | 642 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
