Assay Details
Gene Symbol: aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199690.2 | NP_955984.2 | 8 - 9 | 951 | 67 | |
GenBank mRNA | BC059643.1 | - | 8 - 9 | 949 | 67 | |
BC065447.1 | - | 8 - 9 | 951 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
