Assay Details
Gene Symbol: ldb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199858.3 | NP_956152.1 | 7 - 8 | 882 | 111 | |
XM_005156271.4 | XP_005156328.1 | 6 - 7 | 979 | 111 | ||
XM_005156272.4 | XP_005156329.1 | 8 - 9 | 936 | 111 | ||
XM_005156273.4 | XP_005156330.1 | 8 - 9 | 982 | 111 | ||
XM_009306591.3 | XP_009304866.1 | 6 - 7 | 1025 | 111 | ||
GenBank mRNA | BC047189.1 | - | 8 - 9 | 917 | 111 | |
BC065846.1 | - | 7 - 8 | 887 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
