Assay Details
Gene Symbol: pelo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201136.1 | NP_957430.1 | 6 - 7 | 705 | 120 | |
GenBank mRNA | BC049484.1 | - | 6 - 7 | 705 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
