Assay Details
Gene Symbol: atg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009914.2 | NP_001009914.2 | 2 - 3 | 298 | 120 | |
NM_205618.2 | NP_991181.2 | 2 - 3 | 245 | 120 | ||
GenBank mRNA | AY398310.1 | - | 2 - 3 | 203 | 120 | |
BC077086.1 | - | 2 - 3 | 297 | 120 | ||
HQ450378.1 | - | 1 - 2 | 114 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
