Assay Details
Gene Symbol: atg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009914.2 | NP_001009914.2 | 4 - 5 | 506 | 87 | |
NM_205618.2 | NP_991181.2 | 4 - 5 | 453 | 87 | ||
GenBank mRNA | AY398310.1 | - | 4 - 5 | 411 | 87 | |
BC077086.1 | - | 4 - 5 | 505 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
