Assay Details
Gene Symbol: atg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009914.2 | NP_001009914.2 | 5 - 6 | 669 | 93 | |
NM_205618.2 | NP_991181.2 | 5 - 6 | 616 | 93 | ||
GenBank mRNA | AY398310.1 | - | 5 - 6 | 574 | 93 | |
BC077086.1 | - | 5 - 6 | 668 | 93 | ||
HQ450378.1 | - | 4 - 5 | 485 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
