Assay Details
Gene Symbol: myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212814.2 | NP_997979.2 | 2 - 3 | 560 | 133 | |
GenBank mRNA | AY388401.1 | - | 2 - 3 | 424 | 133 | |
BC083419.1 | - | 2 - 3 | 560 | 133 | ||
BC164642.1 | - | 2 - 3 | 458 | 133 | ||
DQ100359.1 | - | 2 - 3 | 480 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
