Assay Details
Gene Symbol: arhgap11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193539.1 | NP_001180468.1 | 5 - 6 | 1010 | 88 | |
NM_213316.2 | NP_998481.2 | 5 - 6 | 1010 | 88 | ||
XM_005173570.4 | XP_005173627.1 | 5 - 6 | 1408 | 88 | ||
GenBank mRNA | BC066588.1 | - | 5 - 6 | 758 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
