Assay Details
Gene Symbol: aimp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213338.1 | NP_998503.1 | 2 - 3 | 449 | 117 | |
XM_021470105.1 | XP_021325780.1 | 2 - 3 | 384 | 117 | ||
GenBank mRNA | BC053178.1 | - | 2 - 3 | 449 | 117 | |
BC164848.1 | - | 2 - 3 | 379 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
