Assay Details
Gene Symbol: kyat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213359.2 | NP_998524.2 | 12 - 13 | 1344 | 145 | |
XM_005171922.4 | XP_005171979.1 | 12 - 13 | 1444 | 145 | ||
XM_005171923.4 | XP_005171980.1 | 12 - 13 | 1432 | 145 | ||
GenBank mRNA | BC055182.1 | - | 12 - 13 | 1264 | 145 | |
DT059481.1 | - | 5 - 6 | 536 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
