Assay Details
Gene Symbol: aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214712.1 | NP_999877.1 | 3 - 4 | 472 | 109 | |
GenBank mRNA | AF274877.1 | - | 3 - 4 | 472 | 109 | |
BC078231.1 | - | 3 - 4 | 496 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
