Assay Details
Gene Symbol: tuba7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002230.1 | NP_001002230.1 | 2 - 3 | 331 | 86 | |
XM_021479153.1 | XP_021334828.1 | 1 - 2 | 740 | 86 | ||
GenBank mRNA | BC072721.1 | - | 2 - 3 | 331 | 86 | |
BC165338.1 | - | 1 - 2 | 257 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
