Assay Details
Gene Symbol: adprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003453.2 | NP_001003453.1 | 2 - 3 | 408 | 99 | |
GenBank mRNA | BC078399.1 | - | 2 - 3 | 254 | 99 | |
BC164170.1 | - | 1 - 2 | 146 | 99 | ||
EH441896.1 | - | 2 - 3 | 406 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
