Assay Details
Gene Symbol: nck2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003492.2 | NP_001003492.2 | 3 - 4 | 1144 | 99 | |
GenBank mRNA | BC078356.1 | - | 3 - 4 | 1112 | 99 | |
BC096988.1 | - | 3 - 4 | 1144 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
