Assay Details
Gene Symbol: sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003523.2 | NP_001003523.2 | 5 - 6 | 511 | 96 | |
GenBank mRNA | BC078306.1 | - | 5 - 6 | 429 | 96 | |
EH444095.1 | - | 3 - 4 | 307 | 96 | ||
EH540297.1 | - | 5 - 6 | 528 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
