Assay Details
Gene Symbol: ndufs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020676.1 | NP_001018512.1 | 1 - 2 | 90 | 88 | |
GenBank mRNA | BC095734.1 | - | 1 - 2 | 90 | 88 | |
BC155231.1 | - | 1 - 2 | 90 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
