Assay Details
Gene Symbol: atg4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024434.1 | NP_001019605.1 | 2 - 3 | 138 | 69 | |
GenBank mRNA | BC095617.1 | - | 2 - 3 | 138 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
